Learn More
Abeba™ UNI6 36751 Shoes
Product Code. 15690508
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15690508 35 1 pair
15600518 36 1 pair
15610518 37 1 pair
15620518 38 1 pair
15630518 39 1 pair
15640518 40 1 pair
15650518 41 1 pair
15660518 42 1 pair
15670518 43 1 pair
15680518 44 1 pair
15690518 45 1 pair
15600528 46 1 pair
15610528 47 1 pair
15620528 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15690508 Supplier Abeba™ Supplier No. 36751/35
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Smooth leather
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color Black
Liner Material Microfiber
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36751