Learn More
ABEBA™ Uni6 Black Safety Slip-On Shoe S2
Product Code. 16348021
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16348021 45 1 pair
16348011 35 1 pair
16358011 36 1 pair
16368011 37 1 pair
16378011 38 1 pair
16388011 39 1 pair
16398011 40 1 pair
16308021 41 1 pair
16318021 42 1 pair
16328021 43 1 pair
16338021 44 1 pair
16358021 46 1 pair
16368021 47 1 pair
16378021 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16348021 Supplier ABEBA™ Supplier No. 31741/45
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
ABEBA™ Uni6 Black Safety Slip-On Shoe S2 made of water and chemical resistant microfiber with a slip resistant PU/TPU outsole.
  • Comfort, safety and hygiene
  • Comply to EN ISO 20345 : 2012, S2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Water and chemical resistant microfiber
  • Steel toe cap (200 joules)
  • Instep area with elastic band
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • No side stitching
  • Removable insole Active comfort
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2 SRC ESD
Color Black
Liner Material Breathable silver ions lining
Lug Type Padded
Material Microfiber
Size 45
Sole Features Slip and abrasion resistant, ESD dissipative
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Slip-on shoes
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes