Learn More
ABEBA™ Uni6 Black Safety Trainers S3
Product Code. 17191115
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17191115 42 1 pair
17121115 35 1 pair
17131115 36 1 pair
17121125 45 1 pair
17131125 46 1 pair
17141125 47 1 pair
17151125 48 1 pair
17141115 37 1 pair
17151115 38 1 pair
17161115 39 1 pair
17171115 40 1 pair
17181115 41 1 pair
17101125 43 1 pair
17111125 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17191115 Supplier ABEBA™ Supplier No. 1676/42
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of black water repellent and breathable fabric, slip resistant PU/TPU outsole, steel toe cap and textile antipuncture midsole
  • Comfort, safety and style
  • Comply to EN ISO 20345: 2012, S3, SRC
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Removable insole
  • Active comfort
Specifications
Certifications/Compliance EN ISO 20345: 2012, S3, SRC
Color Black
Material Knitted Textile
Size 42
Toe Material Steel
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type S3 SRC