Learn More
ABEBA™ Uni6 White Occupational Slip-On Shoes O2
Product Code. 16398111
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16398111 37 1 pair
16378111 35 1 pair
16388111 36 1 pair
16308121 38 1 pair
16318121 39 1 pair
16328121 40 1 pair
16338121 41 1 pair
16348121 42 1 pair
16358121 43 1 pair
16368121 44 1 pair
16378121 45 1 pair
16388121 46 1 pair
16398121 47 1 pair
16308131 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16398111 Supplier ABEBA™ Supplier No. 36740/37
Only null left
Add to basket
Add to basket
ABEBA™ Uni6 White Occupational Slip-On Shoes O2 made of water and chemical resistant microfiber with a washable, slip resistant PU/TPU outsole.
  • Hygiene, comfort
  • Comply to EN ISO 20347 : 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water and chemical resistant microfiber
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
  • Washable at 30°C
Specifications
Certifications/Compliance CE EN ISO 20347 :2012, O2, FO, SRC, ESD
Color White
Liner Material Breathable silver ions lining
Material Microfiber
Size 37
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Slip-on shoes
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20347 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes