Learn More
ABEBA™ Uni6 White Occupational Sneakers O1
Product Code. 16368141
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 16368141
Brand: ABEBA™ 36772/36
Only null left
Add to basket
Add to basket
ABEBA™ Uni6 White Occupational Sneakers O1 made of water repellent knitted textile with a slip resistant PU/TPU outsole.
  • Breathability, lightweight, comfort
  • Comply to EN ISO 20347 : 2012, O1 FO, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent and breathable knitted textile
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
Specifications
Certifications/Compliance CE EN ISO 20347 :2012, O1, FO, SRC, ESD
Closure Type Laces
Color White
Eyelets Material Stainless steel
Liner Material Breathable silver ions lining
Material Textile
Size 36
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Sneakers
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20347 : 2012; DIN EN 61340