Learn More
ABEBA™ Uni6 White Occupational Sneakers O2
Product Code. 16338101
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16338101 35 1 pair
16348101 36 1 pair
16358101 37 1 pair
16368101 38 1 pair
16378101 39 1 pair
16388101 40 1 pair
16398101 41 1 pair
16308111 42 1 pair
16318111 43 1 pair
16328111 44 1 pair
16338111 45 1 pair
16348111 46 1 pair
16358111 47 1 pair
16368111 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16338101 Supplier ABEBA™ Supplier No. 36729/35
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
ABEBA™ Uni6 White Occupational Sneakers O2 made of tear and water resistant fabric with slip resistant PU/TPU outsole.
  • Lightweight, supple, water resistant
  • Comply to EN ISO 20347 : 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Tear and water resistant fabrics
  • Easy fastening with laces
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
Specifications
Certifications/Compliance CE EN ISO 20347 :2012, O2, FO, SRC, ESD
Closure Type Laces
Color White
Eyelets Material Stainless steel
Liner Material Breathable silver ions lining
Material Textile
Size 35
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Trainer
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20347 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes