Learn More
ABEBA™ Uni6 White Safety Slip-On Shoe S2
Product Code. 16308001
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16308001 35 1 pair
16318001 36 1 pair
16328001 37 1 pair
16338001 38 1 pair
16348001 39 1 pair
16358001 40 1 pair
16368001 41 1 pair
16378001 42 1 pair
16388001 43 1 pair
16398001 44 1 pair
16308011 45 1 pair
16318011 46 1 pair
16328011 47 1 pair
16338011 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16308001 Supplier ABEBA™ Supplier No. 31740/35
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
ABEBA™ Uni6 White Safety Slip-On Shoe S2 made of water and chemical resistant microfiber with a slip resistant PU/TPU outsole.
  • Comfort, safety and hygiene
  • Comply to EN ISO 20345 : 2012, S2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Water and chemical resistant microfiber
  • Steel toe cap (200 joules)
  • Instep area with elastic band
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • No side stitching
  • Removable insole Active comfort
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2 SRC ESD
Color White
Liner Material BreathaBle silver ions lining
Lug Type Padded
Material Microfiber
Size 35
Sole Features Slip and abrasion resistant, ESD dissipative
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Slip-on shoes
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes