Description
UNICORN start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA start instrument, and to evaluate and share results. While ÄKTA start can function as a stand-alone system, UNICORN start software maximizes the performance of the instrument.
- Simple and flexible method creation using predefined templates and phases (steps)
- Intuitive graphic user interface enables users to perform many operations in a single-click
- Enables real-time remote monitoring of the run and an intuitive way to control the run
- Evaluate, compare, and share results; create and print PDF reports
- Robust database storage with easy access to data, data security, and data integrity
Specifications
Specifications
| Type | Version 1.1 |
| Description | System control software |
| For Use With (Equipment) | ÄKTA Start |
| Includes | DVD, activation code |