Learn More
uvex Unidur 6641 Cut Protection Gloves
Product Code. 15346972 Shop All Uvex Products
Change view
Click to view available options
Size:
6
11
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15346972 11 Each
17240411 6 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15346972 Supplier uvex Supplier No. 6021011
  • ONLINE EXCLUSIVE PRICE
    12.05 €
    11.82 € / Pack of 1
    Save 0.23 €
    1% Off
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Gloves are constructed from high quality HPPE fibers, with a polyurethane coating for enhanced grip capabilities and level B cut resistance.
  • Lightweight, polyurethane-coated HPPE assembly gloves
  • Coating provides mechanical abrasion resistance and level 3/B cut protection
  • Enhanced grip in dry or slightly damp areas
  • High level of sensitivity for tasks that require tactility
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color White, Gray
Hand Ambidextrous
Latex Free Yes
Material HPPE, Elastane
Product Type Cut-resistant Gloves
Size 11
EN388 Standard 4343B
Glove Cuff Style Knit
Series Unidur 6641