missing translation for 'onlineSavingsMsg'
Learn More

uvex Unidur 6641 Cut Protection Gloves

Product Code. 17240411 Shop All Uvex Products
Change view
Click to view available options
Size:
6
11
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17240411 6 10 pairs
15346972 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17240411 Supplier uvex Supplier No. 6021006
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Gloves are constructed from high quality HPPE fibers, with a polyurethane coating for enhanced grip capabilities and level B cut resistance.

  • Lightweight, polyurethane-coated HPPE assembly gloves
  • Coating provides mechanical abrasion resistance and level 3/B cut protection
  • Enhanced grip in dry or slightly damp areas
  • High level of sensitivity for tasks that require tactility
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Polyurethane
Color White, Gray
Hand Ambidextrous
Latex Free Yes
Material HPPE, Elastane
Product Type Cut-resistant Gloves
Size 6
EN388 Standard 4343B
For Use With (Application) Automotive, Metal Work, and Packaging
Gender Unisex
Glove Cuff Style Knit
Series Unidur 6641
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.