missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unilite Thermo HD Safety Gloves

Product Code. 17240491 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17240491 11 10 pairs
17210491 8 10 pairs
17220491 9 10 pairs
17230491 10 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17240491 Supplier Uvex™ Supplier No. 6094211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Two-layer acylic and cotton terry construction provides protection in cold environment applications. PVC coating on the palm and back of hand enhances grip capabilities, and remains flexible even at low temperatures.

  • Two-layer acrylic, cotton terry contruction provides reliable protection in cold applications
  • PVC coating on the palm and fingers enhances grip capabilites
  • Gloves remain comfortable and flexible, even at low temperatures
  • Provides thermal insulation when in direct contact with cold objects
  • Mechanical protection for heavy-duty tasks
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 511: 12X
Applications Industrial
Coating Material PVC
Color Orange, Black
Material Acrylic, Terrycloth
Size 11
EN388 Standard 3231X
For Use With (Application) Cold Environments, Construction, Forklift Operation, and Refrigeration
Gender Unisex
Glove Cuff Style Knit
Series Unilite Thermo HD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.