missing translation for 'onlineSavingsMsg'
Learn More

uvex Unilite Thermo Plus Cut C Safety Gloves

Product Code. 17230461 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17230461 7 10 pairs
17240461 8 10 pairs
17250461 9 10 pairs
17260461 10 10 pairs
17270461 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17230461 Supplier uvex Supplier No. 6059107
  • ONLINE EXCLUSIVE PRICE
    140.00 €
    137.45 € / 10 pairs
    Save 2.55 €
    1% Off
View all promotions
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Winter safety gloves combine protection against thermal risks with level C cut protection. Polymer coating enhances grip and remains flexible even at low temperatures.

  • Winter safety gloves combine thermal protection with level C cut protection
  • Robust polymer coating remains flexible even at low temperatures
  • Acrylic liner provides insulation when in direct contact with cold objects
  • Enhanced grip in dry or slightly damp areas
  • Dual-layer construction
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 511: 02X
Applications Industrial
Coating Material Polymer
Color Green, Black
Exterior Finish Smooth
Hand Ambidextrous
Material Polyamide, Glass
Product Type Cold-resistant Gloves
Size 7
EN388 Standard 3X42C
For Use With (Application) Construction, Forklift Operation, Metal Work, and Refrigeration
Gender Unisex
Glove Cuff Style Knit
Liner Material Acrylic
Series Unilite Thermo Plus Cut C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.