Learn More
uvex Unilite Thermo Plus Safety Gloves
Product Code. 15377132 Shop All Uvex Products
Change view
Click to view available options
Size:
7
9
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15377132 9 Each
17280461 7 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15377132 Supplier uvex Supplier No. 6059209
  • ONLINE EXCLUSIVE PRICE
    14.00 €
    9.05 € / 1 pairs
    Save 4.95 €
    35% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Two-layer material composition features acrylic and new wool to ensure a pleasant hand climate and enhanced user comfort. Polymer coating provides mechanical abrasion resistance, and remains flexible even at low temperatures.
  • Gloves feature two-layer material composition of acrylic and new wool
  • Polymer coating on the palm remains flexible even at low temperatures
  • Coating provides mechanical abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp areas
  • Provides thermal insulation when in direct contact with cold objects
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polymer
Color Black
Exterior Finish Smooth
Hand Ambidextrous
Material Polyamide, Elastane
Product Type Cold-resistant Gloves
Size 9
EN388 Standard 3131X
Glove Cuff Style Knit
Liner Material Acrylic, Wool
Series Unilite Thermo Plus