missing translation for 'onlineSavingsMsg'
Learn More

uvex Unilite Thermo Plus Safety Gloves

Product Code. 17280461 Shop All Uvex Products
Change view
Click to view available options
Size:
7
9
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17280461 7 10 pairs
15377132 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17280461 Supplier uvex Supplier No. 6059207
  • ONLINE EXCLUSIVE PRICE
    92.00 €
    81.98 € / 10 pairs
    Save 10.02 €
    10% Off
View all promotions
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Two-layer material composition features acrylic and new wool to ensure a pleasant hand climate and enhanced user comfort. Polymer coating provides mechanical abrasion resistance, and remains flexible even at low temperatures.

  • Gloves feature two-layer material composition of acrylic and new wool
  • Polymer coating on the palm remains flexible even at low temperatures
  • Coating provides mechanical abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp areas
  • Provides thermal insulation when in direct contact with cold objects
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 511: 010
Applications Industrial
Coating Material Polymer
Color Black
Exterior Finish Smooth
Hand Ambidextrous
Material Polyamide, Elastane
Product Type Cold-resistant Gloves
Size 7
EN388 Standard 3131X
For Use With (Application) Construction, Forklift Operation, and Refrigeration
Gender Unisex
Glove Cuff Style Knit
Liner Material Acrylic, Wool
Series Unilite Thermo Plus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.