missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Universal Foam Brush Set

Product Code. 12353810 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12353810 White Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12353810 Supplier Cole-Parmer™ Supplier No. 8455200
  • ONLINE EXCLUSIVE PRICE
    55.25 €
    54.68 € / Pack of 3
    Save 0.57 €
    1% Off
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clean a variety of interior surfaces without absorbing water. Universal Foam Brush Set includes 9 in. and 18 in. length washing brushes and a 15 in. length universal glassware brush. Long-lasting brushes last up to 20 times longer than sponges.

  • Soft, plush foam that gently cleans interior surfaces without absorbing water, meaning foam bristles never become soggy or limp.
  • These long lasting brushes last up to ten times longer than top quality brushes and up to twenty times longer than sponges.
  • The three brush set includes 229 mm and 457 mm long washing brushes and a 381 mm long universal glassware brush.
  • The washing brushes fit glassware with a 63.5 mm width or larger neck.
  • The universal brush measures 19 mm in diameter and bends to conform to the shape of the glassware.
  • When bent and rotated, it cleans corners of the glass that would otherwise be unreachable.

Specifications

Product Type Beaker/Bottle Brush
Bristle Material Foam
Bristle Type Flexible
Color-coded No
Form Foam
Sterility Non-sterile
Color White
For Use With (Equipment) Glassware
Size Universal, 9 in., and 18 in. Long
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.