Learn More
Eppendorf™ Universal Buckets for S-4xUniversal Rotor
Product Code. 15893212 Shop All Eppendorf Products
Change view
Click to view available options
Quantity:
2 Pk.
4 Pk.
Unit Size:
Pack of 2
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15893212 4 Pk. Pack of 4
15883212 2 Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15893212 Supplier Eppendorf™ Supplier No. 5895203000
  • ONLINE EXCLUSIVE PRICE
    2245.00 €
    2199.00 € / Pack of 4
    Save 46.00 €
    2% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Universal Buckets for S-4xUniversal Rotor are suitable swing-buckets for the S-4xUniversal Rotor, accepting a variety of adapters for tubes, flasks, and plates.
  • Suitable for S-4xUniversal Rotor
  • Accepts adapters for tubes, flasks, and plates
  • Variable places depending on adapter used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S-4xUniversal Rotor
Quantity 4 Pk.