missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Universal Clamp with Cork-lined Jaws

Product Code. 10149830
Change view
Click to view available options
Material:
Zinc Die Cast, Powder Coated
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10149830 Zinc Die Cast, Powder Coated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10149830 Supplier Usbeck Laborgeraete™ Supplier No. 2500
  • ONLINE EXCLUSIVE PRICE
    15.55 €
    14.11 € / Pack of 1
    Save 1.44 €
    9% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Secure delicate glassware in the laboratory with the Usbeck Laborgeraete™ Universal Clamp with Cork-lined Jaws.The double-sided opening mechanism allows the jaws to spread evenly for stabilization and balance.

  • Zinc diecasting
  • Capacity 0 – 80 mm
  • Rod Ø 10 mm
  • Powdercoated

Specifications

Type Universal Clamp
Material Zinc Die Cast, Powder Coated
Includes Cork-lined jaws
Diameter (Metric) 0 to 80 mm
Coating Powder Coated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.