Learn More
BOLA Universal Couplings
Click to view available options
Thread Size:
M 6
M 8
Unit Size:
Each
Description
- Coupling made of PTFE for transition from M6 thread to UNF ¼ in. 28 G thread.
- The M6 thread is marked by a groove.
Specifications
Specifications
| Diameter (Metric) Outer | 0.8 mm |
| Thread Size | M 8 |
| Autoclavable | Autoclavable |