missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Universal Human Reference RNA for QuantiGene Assays

Product Code. 32044657
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32044657 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32044657 Supplier Invitrogen™ Supplier No. QS0639
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Universal Human Reference RNA is an RNA extract used as a positive control in QuantiGene assays.

Universal Human Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Human Reference RNA is composed of total RNA from 10 human cell lines:

  • Adenocarcinoma, mammary gland
  • Melanoma
  • Hepatoblastoma, liver
  • Liposarcoma
  • Adenocarcinoma, cervix
  • Histiocytic lymphoma; macrophage; histocyte
  • Embryonal carcinoma, testis
  • Lymphoblastic leukemia, T lymphoblast
  • Glioblastoma, brain
  • Plasmacytoma; myeloma; B lymphocyte

Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Reference RNA
Quantity 10 μg
Sample Source Cell Line
Shipping Condition Shipped at 2°C to 8°C

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.