Learn More
Kartell™ Universal Safety Pipette Bulb
Reduce risk of cracking or chipping when filling any size pipette. Kartell™ Universal Safety Pipette Bulb uses an attchment with an adapter to seal around pipettes without twisting.
Brand: Kartell™ 0020800
Code : NEW
Additional Details : Weight : 0.22500kg
Description
Reduce risk of cracking or chipping when filling any size pipette. Kartell™ Universal Safety Pipette Bulb uses an attchment with an adapter to seal around pipettes without twisting.
- Red rubber with silicon bulb
- For use with pipettes with a capacity of up to 25mL
Specifications
Rubber and Silicone | |
Universal Pipette Filler |