missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Universal Shoecovers with Printed Chevron Sole

Product Code. 17720159 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17720159 Blue Pack of 500
17710159 White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17720159 Supplier Kolmi Hopen™ Supplier No. H10020
  • ONLINE EXCLUSIVE PRICE
    83.00 €
    67.15 € / Pack of 500
    Save 15.85 €
    19% Off
View all promotions
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made of polypropylene with a latex sole, these shoecovers provide the slip-resistance you need when walking on smooth floors.

  • Sole with 'herringbone' print
  • Elastic ankles
  • Polypropylene with latex sole
  • Perfectly adapted to smooth surfaces in dry environments

Specifications

Size Universal
Material Polypropylene, Latex
Latex Free No
Color Blue
Disposable Single-use
For Use With (Application) Industrial
Certifications/Compliance ISO 9001
Packaging Quantity 500 Pk.
Dimensions (L x W) 400 x 170 mm
Weight (Metric) 40 g/m2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.