missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Unmodified Cellulose MN 100

Brand:  Macherey-Nagel™ 815050.1

Code : NEW

Additional Details : CAS Number : 7631-86-9 Weight : 1.00000kg

 View more versions of this product

Product Code. 10033081

  • 26.70 € / 1kg

Estimated Shipment: 11-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Standard
1 kg
∼6500 cm2/g
Unmodified Cellulose MN 100
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers