Learn More
Kartell™ Plastilab Untapered T-Shaped Tubing Connectors
Product Code. 12573297
Change view
Click to view available options
Port Size:
1.6mm bore
3.7mm
4.4mm bore
6.3mm bore
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Port Size Packaging Quantity Unit Size
12573297 3.7mm 100/Pk. Pack of 100
12569746 1.6mm bore 10/Pk. Pack of 10
11904359 4.4mm bore 10/Pk. Pack of 10
12090582 6.3mm bore 10/Pk. Pack of 10
12519666 4.4mm bore 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12573297 Supplier Kartell™ Supplier No. 0046000
  • ONLINE EXCLUSIVE PRICE
    67.00 €
    43.86 € / Pack of 100
    Save 23.14 €
    34% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations
Specifications
Certifications/Compliance Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Suitable for foodstuff
Packaging Quantity 100/Pk.
Port Size 3.7mm
Shape T-shape
Product Type Untapered T Tubing Connector