Learn More
R&D Systems™ UNveil Unstained Protein Ladder
Product Code. 30175121
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Product Code. Quantity Unit Size
30175121 500 μL 500µL
1 options
Product Code. 30175121 Supplier R&D Systems™ Supplier No. NBP333175
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Unstained Protein Ladder
The Unveil Unstained Protein Ladder is a mixture of 12 unstained recombinant proteins covering a wide range of molecular weights from 10 to 200 kDa. The proteins resolve into clearly identifiable bands when separated on the SDS-PAGE gel (MES buffer), with the intensified 25kDa and 85kDa protein bands serving as the two reference bands. The Unveil Unstained Protein Ladder is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis and verifying Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins. The ladder is supplied in the gel loading buffer and is ready to use.
TRUSTED_SUSTAINABILITY
Specifications
Formulation 20 mM Tris-phosphate (pH 7.5), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15 % (v/v) Glycerol.
For Use With (Application) 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring
Quantity 500 μL
Product Line Novus