missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals uPA Inhibitor II, UK122

Product Code. 15496619
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496619 25 mg 25mg
15486619 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496619 Supplier Thermo Scientific Chemicals Supplier No. J64874.MB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C17H13N3O2·CF3CO2H

A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activatoruPA Inhibitor II, UK122 acts as a potent, selective and active-site targeting urokinase-type plasminogen activator (uPA) inhibitor. However, its activity against trypsin, thrombin, tPA, and plasmin is greatly reduced. It is also effectively inhibits the migration (80% inhibition) and invasion (68% inhibition) of uPA-expressing CFPAC-1 pancreatic cells, without affecting its proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
Molecular Formula C17H13N3O2·CF3CO2H
Formula Weight 405.30g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.