missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Back Pressure Regulators: Cartridge Holders

Product Code. 12967669
Change view
Click to view available options
For Use With:
Back Pressure Regulator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
12967669 Back Pressure Regulator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12967669 Supplier Kinesis™ Supplier No. 0201709
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    237.32 € / Pack of 1
    Save 28.68 €
    10% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Upchurch Scientific™ Back Pressure Regulators

100% Biocompatible, inert materials.

Specifications

Type Cartridge Holder
Color Black
Description Thread port configuration: 1/4 to 28 flat bottom
For Use With Back Pressure Regulator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.