missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Flangeless Fittings: Ferrules, Tefzel

Product Code. 12997609
Change view
Click to view available options
Color:
Natural
Yellow
For Use With (Equipment):
1/16 in. OD Tubing
1/16 in. OD Tubing, Male Nuts
1/8 in. O.D. Tubing
1/8 in. OD Tubing, Male Nuts
Length (English):
0.16 in.
0.22 in.
Not Applicable
Width (English):
0.18 in.
0.2 in.
Not Applicable
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Length (English) Width (English) Unit Size
12997609 Yellow 1/8 in. OD Tubing, Male Nuts Not Applicable Not Applicable Pack of 10
12917579 Natural 1/16 in. OD Tubing, Male Nuts 0.22 in. 0.18 in. Pack of 10
12957639 Yellow 1/8 in. O.D. Tubing 0.16 in. 0.2 in. Pack of 10
12987609 Natural 1/8 in. OD Tubing, Male Nuts Not Applicable Not Applicable Pack of 10
12987589 Yellow 1/16 in. OD Tubing Not Applicable Not Applicable Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12997609 Supplier Kinesis™ Supplier No. P300X
  • ONLINE EXCLUSIVE PRICE
    35.00 €
    28.97 € / Pack of 10
    Save 6.03 €
    17% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

No need to flange tubing

Excellent for connecting rigid or semi-rigid tubing

Specifications

Type Flangeless Ferrule
Length (English) Not Applicable
Width (English) Not Applicable
Material ETFE
Color Yellow
For Use With (Equipment) 1/8 in. OD Tubing, Male Nuts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.