Learn More
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Popular threaded coned port configurations support quality high-pressure connections
Brand:  Idex F-330X
Product Code. 13033120
  • 134.00 € / Pack of 10
Estimated Shipment: 17-11-2026
Add to basket
Description
Description
Popular threaded coned port configurations support quality high-pressure connections
  • Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Specifications
Specifications
Fingertight Two-Piece Fitting
PEEK
1/16 in. OD Tubing
Ferrule
Natural