missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ LiteTouch™ Ferrule, Natural PEEK/SS, 1/16 in. OD Tubing, 10-32 Coned

Product Code. 13024150
Change view
Click to view available options
For Use With (Equipment):
LiteTouch Ferrule System
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13024150 LiteTouch Ferrule System Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13024150 Supplier Kinesis™ Supplier No. 0193934
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Secure a leak-tight connection.

Specifications

Type Ferrule
Color Natural
Description Fits over 1/16 in. O.D. tubing
For Use With (Equipment) LiteTouch Ferrule System
Material PEEK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.