Learn More
Kinesis™ LuerTight™ Fittings: PEEK/Polypropylene
Product Code. 12997749
Change view
Click to view available options
Material:
PEEK
Unit Size:
Each
Product Code. Material Unit Size
12997749 PEEK Each
1 options
Product Code. 12997749 Supplier Kinesis™ Supplier No. 0201412
  • ONLINE EXCLUSIVE PRICE
    38.00 €
    30.50 € / Pack of 1
    Save 7.50 €
    19% Off
View all promotions
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
  • Male Luer Lock x Female 1/4-28 Flat Bottom
  • Lock hub securely holds female luer adapters in place
  • 0.040 in. thru-hole diameter
  • Manufactured from PEEK
Specifications
Type Luer Adapter
Material PEEK
Color Red