missing translation for 'onlineSavingsMsg'
Learn More

Idex LuerTight™ Fittings: Polypropylene Body

Product Code. 12927759
Change view
Click to view available options
Material:
PEEK
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12927759 PEEK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12927759 Supplier Idex Supplier No. 0201414
  • ONLINE EXCLUSIVE PRICE
    33.00 €
    28.14 € / Pack of 1
    Save 4.86 €
    14% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenience of quick connect and disconnect of a luer connection

  • Manufactured from inert polymers
  • Female Luer x Female 1/4-28 Flat Bottom
  • 0.050 in. thru-hole diameter
  • Manufactured from red PEEK

Specifications

Type Threaded Luer Adapter
Material PEEK
Color Red
For Use With (Equipment) Female Luer x Female 1/4-28 Flat Bottom
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.