missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Plastic Tubing Cutter

Product Code. 12986259
Change view
Click to view available options
Color:
Silver
Material:
Steel
Product Type:
Replacement Blade
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Product Type Unit Size
12986259 Silver Steel Replacement Blade Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12986259 Supplier Kinesis™ Supplier No. 0201686
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing

  • Built-in guide holes for both 1/16 and 1/8 in. OD tubing
  • Replacement blades included and sold separately
Flat, 90°, burr-free cut
Offers durability, reliability and ease
  • Able to cut through polymers such as PEEK™, FEP, PFA and Tefzel™
  • Specifications

    Material Steel
    For Use With Upchurch™ standard polymer tubing cutter
    Color Silver
    Product Type Replacement Blade
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.