missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Precolumn Inline Filters

Protect larger columns and injections from harmful particulates

Brand:  Kinesis™ 02011-01

Code : Z3

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 12946259

  • 150.00 € / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Ideal for higher-flow systems
  • Interchangeable frits make protection more economical
  • Compatible with SFC and HPLC
  • Available to fit a variety of tubing sizes
Specifications

Specifications

0.5 μm
Precolumn Inline Filter
0.94 in.
Stainless Steel
0.66 uL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers