missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Precolumn Inline Filters
Protect larger columns and injections from harmful particulates
92.22 € - 153.00 €
Product Code | Brand | Pore Size | Housing Material | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Housing Material | Volume (Metric) | Price | Quantity & Availability | |||
12936259
|
Idex
02011-00 |
2 μm | Stainless Steel | 0.66 uL |
126.00 €
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
12946259
|
Kinesis™
02011-01 |
0.5 μm | Stainless Steel | 0.66 uL |
150.00 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
12936279
|
Kinesis™
02010-35 |
0.5 μm | PEEK | 1.3 uL |
153.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11574835
|
Idex
A-318 |
0.5 μm | Stainless Steel | 0.23 uL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Ideal for higher-flow systems
- Interchangeable frits make protection more economical
- Compatible with SFC and HPLC
- Available to fit a variety of tubing sizes
Specifications
Precolumn Inline Filter |