missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Quick Connect Adapter

Product Code. 12937759
Change view
Click to view available options
Type:
Female Luer Adapter
Male Luer Adapter
Material:
ETFE
PEEK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material Unit Size
12937759 Female Luer Adapter PEEK Each
12967769 Male Luer Adapter ETFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12937759 Supplier Kinesis™ Supplier No. 0201415
  • ONLINE EXCLUSIVE PRICE
    35.00 €
    28.14 € / Pack of 1
    Save 6.86 €
    19% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenience of quick connect and disconnect of a luer connection

Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.

Specifications

Type Female Luer Adapter
Diameter (English) Inner 0.05 in.
Material PEEK
Color Tan
For Use With (Equipment) Female Luer x Female 10-32 Coned
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.