missing translation for 'onlineSavingsMsg'
Learn More

Idex Stainless-Steel Sample Loop

Product Code. 12963070
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12963070 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12963070 Supplier Idex Supplier No. 0201632
  • ONLINE EXCLUSIVE PRICE
    491.00 €
    418.55 € / Pack of 1
    Save 72.45 €
    14% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compatible with Rheodyne™ analytical injection valves

Leads are long enough to be used at 40° and 60°. For Rheodyne Models 7125, 7010 (leads angled at 40°); and Models 7725, 7725i (leads angled at 60°).

  • Fits into Rheodyne analytical sample injection valves, with leads conveniently bent at appropriate angles to allow trouble-free installation
  • For use with Rheodyne valve models 7125, 7010
  • Theoretical volumes are consistently controlled with use of electrochemical cutting machine (ECM)
  • Each loop passivated and flushed with reagent-grade methanol
  • Fittings and adapters accompanying sample loops rated to 1000psi (69 bar)
  • Volumes stated at ±10% with exact calibration

Each calibrated sample loop is documented and supplied with calibration certificate.

Specifications

Type Sample Loop
Material Stainless Steel
For Use With (Application) Analytical Instrumentation, in-Vitro Diagnostics, Biotechnology, Laboratory
Diameter (English) Inner 0.08 in.
Diameter (Metric) Inner 0.2 cm
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.