missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
Specifications
| Type | Tubing |
| Diameter (English) Inner | 1/16 in. |
| Diameter (English) Outer | 0.040 in. |
| Diameter (Metric) Inner | 1.0 mm |
| Overall Length (English) | 2 in. |
| Overall Length (Metric) | 5 cm |
| Material | Stainless Steel (316) |
| Maximum Pressure | 8100 psi (558 bar) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?