missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID

Product Code. 13080850 Shop All Thermo Scientific Products
Change view
Click to view available options
Overall Length (Metric):
5 cm
10 cm
20 cm
30 cm
Diameter (Metric) Inner:
0.125 mm
0.175 mm
0.25 mm
0.5 mm
1.0 mm
Material:
Stainless Steel (304)
Stainless Steel (316)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Diameter (Metric) Inner Material Unit Size
13080850 5 cm 0.25 mm Stainless Steel (316) Each
10752527 20 cm 0.175 mm Stainless Steel (316) Each
13000910 5 cm 1.0 mm Stainless Steel (316) Each
13000850 5 cm 0.5 mm Stainless Steel (304) Each
13020850 20 cm 0.5 mm Stainless Steel (304) Each
13050920 10 cm 0.125 mm Stainless Steel (316) Each
13070920 30 cm 0.125 mm Stainless Steel (316) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 13080850 Supplier Kinesis™ Supplier No. 0201090
  • ONLINE EXCLUSIVE PRICE
    11.00 €
    8.84 € / Pack of 1
    Save 2.16 €
    19% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Type Tubing
Color Blue
Diameter (English) Inner 1/16 in.
Diameter (English) Outer 0.010 in.
Diameter (Metric) Inner 0.25 mm
Overall Length (English) 2 in.
Overall Length (Metric) 5 cm
Material Stainless Steel (316)
Maximum Pressure 19,700 psi (1358 bar)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.