Learn More
Kinesis™ Threaded Plugs and Caps
Product Code. 12937619
Change view
Click to view available options
Type:
Plug
Color:
Natural
White
Material:
Delrin
PFA
Tefzel
Port Type:
1/4-28 Flat Bottom
6-32 Coned
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Color Material Port Type unitSize
12937619 Plug Natural Tefzel 1/4-28 Flat Bottom Each
16593232 Plug White Delrin 6-32 Coned Each
12967619 Plug Natural PFA 1/4-28 Flat Bottom Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12937619 Supplier Kinesis™ Supplier No. 0201378
In Stock
Add to basket
Add to basket
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
Type Plug
Color Natural
Port Type 1/4-28 Flat Bottom
Material Tefzel