Learn More
Syngene™ Upgrade GBOX-F3 to XRQ Camera Kit
GREENER_CHOICE
Product Code. 15869972 Shop All Syngene Products
Click to view available options
For Use With (Equipment):
Gbox F3
Unit Size:
Each
Product Code. 15869972
Brand: Syngene™ 9130400200
Only null left
Add to basket
Add to basket
Syngene™ Upgrade GBOX-F3 to XRQ Camera Kit is required to upgrade existing Gbox F3 systems to Gbox Chemi-XRQ
Specifications
For Use With (Equipment) Gbox F3
Quantity 1
Product Type Upgrade Kit