missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Upper Basket, Height Adjustable

Product Code. 15415464 Shop All Miele Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15415464 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415464 Supplier Miele Professional™ Supplier No. 12645600
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Upper basket, height adjustable, for the optimum loading of inserts

  • Width 475 mm, depth 443 mm
  • With integrated spray arm
TRUSTED_SUSTAINABILITY

Specifications

Color Stainless Steel
Material Stainless Steel
For Use With (Equipment) PG 8583/PG8593
Depth (Metric) 527 mm
Product Type Upper basket
Height (Metric) 206 mm
Width (Metric) 528 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.