Learn More
Berthold Technologies Upper Part For DeWit Tubes
Product Code. 31777359 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777359 Imaging System Each
1 options
Product Code. 31777359 Supplier Berthold Technologies Supplier No. 5659903
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Upper part for the turn table in NightSHADE imaging systems to hold 18 DeWit tubes for sequential imaging.
  • The turntable for NightSHADE enables sequential and scheduled acquisition of up to 18 DeWitt tubes (in groups of three)
  • The component represents the upper part of the turntable.
  • Used together with the turntable base
Specifications
Type Specimen Tray
For Use With (Application) Imaging of Seedlings
For Use With (Equipment) Imaging System