missing translation for 'onlineSavingsMsg'
Learn More

Uracil-5-boronic acid, 95%

Product Code. 15433637
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433637 250 mg 250mg
11352209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433637 Supplier Thermo Scientific Alfa Aesar Supplier No. H27219.MD
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70523-22-7 | C4H5BN2O4 | 155.90 g/mol

It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70523-22-7
Molecular Formula C4H5BN2O4
Molecular Weight (g/mol) 155.90
MDL Number MFCD01318983
InChI Key PVEJOCQTIVCDNO-UHFFFAOYSA-N
Synonym uracil-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl boronic acid, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 2,4-dihydroxypyrimidine-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-ylboronic acid, 2,4-dihydroxypyrimidin-5-ylboronic acid, 2,4-dioxo-1h-pyrimidin-5-yl boronic acid, 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid, t6mvmvj ebqq wln, 5-dihydroxyboryluracil
PubChem CID 256205
IUPAC Name (2,4-dioxo-1H-pyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 155.9
Percent Purity 95%
Chemical Name or Material Uracil-5-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.