missing translation for 'onlineSavingsMsg'
Learn More

Uracil, Extra Pure, SLR, Fisher Chemical

Product Code. 10599200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10599200 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10599200 Supplier Fisher Chemical Supplier No. U/0030/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 66-22-8
Melting Point 335°C
Color White
Packaging Plastic powder jar
Molecular Formula C4H4N2O2
MDL Number MFCD00006016
Quantity 25 g
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILES O=C1NC=CC(=O)N1
IUPAC Name 1H-pyrimidine-2,4-dione
Molecular Weight (g/mol) 112.09
PubChem CID 1174
ChEBI CHEBI:17568
Formula Weight 112.09g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.