missing translation for 'onlineSavingsMsg'
Learn More

Uracil, TRC

Product Code. 17991795
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991795 1 g 1g
17900885 5 g 5g
17950435 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991795 Supplier Toronto Research Chemicals Supplier No. TRCU8010001G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4 H4 N2 O2
Molecular Weight (g/mol) 112.09
Synonym Uracil (8CI), 2,4-Dihydroxypyrimidine, 2,4-Dioxopyrimidine, 2,4-Pyrimidinediol, 2,4-Pyrimidinedione, 4-Hydroxyuracil, Hybar X, NSC 3970, Pirod, Pyrod, 2,4(1H,3H)-Pyrimidinedione, Pyrimidine-2,4(1H,3H)-dione, Uracil, Lamivudine Imp. F (EP), Fluorouracil Imp. C (EP)
IUPAC Name 1H-pyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)
Formula Weight 112.0273
Percent Purity >95%
Chemical Name or Material Uracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.