missing translation for 'onlineSavingsMsg'
Learn More

Urea, MP Biomedicals™

Product Code. 11461702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 lb.
Unit Size:
1lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461702 1 lb. 1lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461702 Supplier MP Biomedicals Supplier No. 04821519
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57-13-6
Melting Point 133°C
Molecular Formula CH4N2O
Quantity 1 lb.
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
Solubility Information Insoluble in Alcohol
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Molecular Weight (g/mol) 60.06
PubChem CID 1176
ChEBI CHEBI:48376
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.