missing translation for 'onlineSavingsMsg'
Learn More

Urea, 98%, extra pure, pearls

Product Code. 10648124 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10648124 10 kg Plastic drum 10kg
10434795 1 kg Plastic Bottle 1kg
10266970 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10648124 Supplier Thermo Scientific Acros Supplier No. 197460100
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex N)
Packaging Plastic drum
Linear Formula H2NCONH2
Quantity 10 kg
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL
Formula Weight 60.06
Percent Purity 98%
Grade Extra Pure
Physical Form Pearls
Chemical Name or Material Urea, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.