missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99.3+%

Product Code. 11367898
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367898 2.5 kg 2.5kg
11357898 100 g 100g
11377898 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367898 Supplier Thermo Scientific Alfa Aesar Supplier No. 036429.A4
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-13-6 | CH4N2O | 60.056 g/mol

Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Color White
Density 1.335 g/mL
pH 9.2 to 9.5
Odor Ammonia-like
Linear Formula NH2CONH2
Quantity 2.5 kg
Beilstein 635724
Merck Index 14,9867
Solubility Information Soluble in water,glycerol and ethanol.
Formula Weight 60.06
Percent Purity ≥99.3%
Physical Form Crystalline Solid
Chemical Name or Material Urea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.