missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥99.5%, Molecular Biology Grade, Ultrapure

Product Code. 15665327 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15665327 5 kg Plastic bottle 5kg
15845488 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15665327 Supplier Thermo Scientific Supplier No. J75826.A7
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chaotropic agent used in denaturing polyacrylamide gel electrophoresis. When 8M urea is not sufficiently denaturing, 40% formamide may be included. | CAS: 57-13-6 | CH4N2O | 60.056 g/mol

  • Chaotropic agent used in denaturing polyacrylamide gel electrophoresis.
  • When 8M urea is not sufficiently denaturing, 40% formamide may be included.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Density 1.335
Packaging Plastic bottle
Linear Formula NH2CONH2
Quantity 5 kg
Beilstein 635724
Specific Gravity 1.335
Formula Weight 60.06
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Urea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.