missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urea, 99+%, for biochemistry

Product Code. 11926921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11926921 5 kg Plastic drum 5kg
11916921 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926921 Supplier Thermo Scientific Chemicals Supplier No. 434720050
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131.0°C to 135.0°C
Color White
pH 7.5 to 9.5
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula H2NCONH2
Merck Index 15,10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL
Formula Weight 60.06
Percent Purity ≥99%
Grade Biochemical
Infrared Spectrum Authentic
Loss on Drying 0.1% max.
Packaging Plastic drum
Residue after Ignition 0.1% max
Chemical Name or Material Urea, For Biochemistry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.